Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mucl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009268.1 | NP_033294.1 | 463 | |||
GenBank mRNA | BC012233.1 | - | 453 | 4165141 | ||
BC024641.1 | - | 454 | 4165428 | |||
BC059095.1 | - | 463 | 4925159 | |||
BC091766.1 | - | 395 | 4190032 | |||
BC093507.1 | - | 463 | 4921535 | |||
M33975.1 | - | 463 |