Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009281.3 | NP_033307.2 | 6 | 485 | ||
GenBank mRNA | AF011758.2 | - | 434 | |||
AK028056.1 | - | 6 | 478 | |||
AK029078.1 | - | 6 | 477 | |||
AK029926.1 | - | 6 | 750 | |||
BC037658.1 | - | 5 | 395 | 5356405 |