Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stag1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009282.3 | NP_033308.2 | 9 | 1134 | ||
GenBank mRNA | AK048671.1 | - | 9 | 1310 | ||
AK052003.1 | - | 9 | 1142 | |||
BC062954.1 | - | 9 | 1134 | 30537388 | ||
BC069841.1 | - | 9 | 989 | |||
Z75332.1 | - | 9 | 1183 |