Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stat6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009284.2 | NP_033310.2 | 22 | 2739 | ||
GenBank mRNA | AF481808.1 | - | 21 | 2454 | ||
AF481809.1 | - | 21 | 2454 | |||
AK135911.1 | - | 5 | 502 | |||
AK149563.1 | - | 22 | 2700 | |||
AK163053.1 | - | 22 | 2740 | |||
BC029318.1 | - | 9 | 928 | |||
BC127054.1 | - | 22 | 2529 | 40109712 |