Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111296.2 | NP_001104766.1 | 2 | 332 | ||
GenBank mRNA | BC145167.1 | - | 2 | 304 | ||
L02335.1 | - | 2 | 305 |
Gene Symbol: Sult2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009286.2 | NP_033312.2 | 2 | 331 | ||
GenBank mRNA | AK149424.1 | - | 2 | 332 | ||
BC125365.1 | - | 304 | 40129880 | |||
BC125367.1 | - | 304 | 40129882 | |||
L27121.1 | - | 304 |