Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111296.2 | NP_001104766.1 | 6 | 855 | ||
GenBank mRNA | BC145167.1 | - | 6 | 827 | ||
L02335.1 | - | 6 | 828 |
Gene Symbol: Sult2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009286.2 | NP_033312.2 | 4 | 581 | ||
GenBank mRNA | AK149424.1 | - | 4 | 582 | ||
BC125365.1 | - | 827 | 40129880 | |||
BC125367.1 | - | 827 | 40129882 | |||
L27121.1 | - | 827 |
Gene Symbol: Sult2a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101534.1 | NP_001095004.1 | 6 | 798 |
Gene Symbol: Sult2a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184980.1 | NP_001171909.1 | 6 | 879 |