Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009288.2 | NP_033314.2 | 9 | 1579 | ||
GenBank mRNA | AK148118.1 | - | 9 | 1522 | ||
AK220448.1 | - | 9 | 1501 | |||
BC069840.1 | - | 9 | 1652 | |||
BC096624.1 | - | 9 | 1664 | |||
BC128363.1 | - | 9 | 1465 | 40111177 | ||
D89728.1 | - | 9 | 1540 |