Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146218.1 | NP_001139690.1 | 5 | 546 | ||
NM_004627.4 | NP_004618.2 | 5 | 656 | |||
GenBank mRNA | AK299144.1 | - | 5 | 499 | ||
AK310268.1 | - | 5 | 1160 | |||
BC012415.1 | - | 5 | 589 | 3865141 | ||
BC111393.1 | - | 5 | 545 | |||
Y12478.1 | - | 5 | 600 |