Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stra8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009292.1 | NP_033318.1 | 3 | 307 | ||
GenBank mRNA | AK006887.1 | - | 4 | 651 | ||
BC100746.1 | - | 2 | 55 | |||
BC103590.1 | - | 3 | 253 | 40045824 | ||
BC103591.1 | - | 3 | 253 | 40045823 | ||
BC103603.1 | - | 3 | 250 | |||
Z75287.1 | - | 3 | 307 |