Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FANCG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004629.1 | NP_004620.1 | 5 | 1067 | ||
GenBank mRNA | AJ007669.1 | - | 5 | 1067 | ||
AK293427.1 | - | 6 | 757 | |||
AK311348.1 | - | 4 | 541 | |||
AK312987.1 | - | 5 | 992 | |||
BC000032.2 | - | 5 | 713 | 3507258 | ||
BC011623.2 | - | 5 | 840 | 4053144 | ||
BT009813.1 | - | 5 | 575 | |||
KU159422.1 | - | 5 | 1098 | |||
KU178053.1 | - | 5 | 575 | |||
KU178054.1 | - | 5 | 575 | |||
KU178055.1 | - | 5 | 575 | |||
KU178056.1 | - | 2 | 149 | |||
U70310.1 | - | 5 | 709 |