Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009305.2 | NP_033331.2 | 7 | 2031 | ||
GenBank mRNA | AK090259.1 | - | 7 | 1933 | ||
BC014823.1 | - | 7 | 1901 | 4507116 | ||
BC052354.1 | - | 7 | 1925 | |||
X95818.1 | - | 7 | 1899 |