Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009308.3 | NP_033334.2 | 2 | 822 | ||
GenBank mRNA | AK032106.1 | - | 2 | 814 | ||
AK043985.1 | - | 2 | 804 | |||
AK050515.1 | - | 2 | 767 | |||
AK083960.1 | - | 2 | 809 | |||
AK148681.1 | - | 2 | 799 | |||
AK184570.1 | - | 1 | 138 | |||
AK220264.1 | - | 2 | 797 | |||
BC052738.1 | - | 750 | ||||
BC058208.1 | - | 2 | 750 | 5361684 |