Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: T | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009309.2 | NP_033335.1 | 8 | 1535 | ||
GenBank mRNA | X51683.1 | - | 8 | 1535 |