Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pou6f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010127.3 | NP_034257.1 | 6 | 2102 | ||
GenBank mRNA | AK031894.1 | - | 1 | 68 | ||
BC053046.1 | - | 8 | 1932 | |||
BC059830.1 | - | 6 | 2102 | 6410550 | ||
BC085139.1 | - | 11 | 2480 | 6406821 |