Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phex | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011077.2 | NP_035207.1 | 3 | 807 | ||
GenBank mRNA | AK158481.1 | - | 3 | 834 | ||
BC137925.1 | - | 3 | 554 | 8860944 | ||
BC137926.1 | - | 3 | 554 | 8860945 | ||
EF194891.1 | - | 3 | 341 | |||
U49908.1 | - | 3 | 799 | |||
U73910.1 | - | 3 | 807 | |||
U75646.1 | - | 3 | 388 |