Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3c2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011083.2 | NP_035213.2 | 2 | 1257 | ||
GenBank mRNA | AK049609.1 | - | 2 | 1247 | ||
AK140908.1 | - | 3 | 1337 | |||
AK142284.1 | - | 3 | 1286 | |||
AK163446.1 | - | 2 | 1240 | |||
BC096553.1 | - | 1 | 691 | |||
U52193.1 | - | 3 | 1163 | |||
U55772.1 | - | 1 | 573 |