Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pikfyve | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011086.2 | NP_035216.2 | 12, 13 | 1654 | ||
GenBank mRNA | AF102777.1 | - | 12, 13 | 1634 |