Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pitx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042502.2 | NP_001035967.1 | 3 | 2050 | ||
NM_001042504.2 | NP_001035969.1 | 4 | 1541 | |||
NM_001286942.1 | NP_001273871.1 | 5 | 1640 | |||
NM_001287048.1 | NP_001273977.1 | 3 | 2050 | |||
NM_011098.4 | NP_035228.2 | 5 | 1679 | |||
GenBank mRNA | AF048723.1 | - | 4 | 1477 | ||
AF048724.1 | - | 3 | 2010 | |||
AF201091.1 | - | 3 | 1377 | |||
BC075660.1 | - | 3 | 1715 | 30606653 | ||
U70132.1 | - | 2 | 1200 | |||
U80010.1 | - | 5 | 1614 | |||
U80011.1 | - | 4 | 1494 | |||
U80036.1 | - | 4 | 1512 |