Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP3B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278511.1 | NP_001265440.1 | 5 | 654 | ||
NM_001278512.1 | NP_001265441.1 | 6 | 750 | |||
NM_004644.4 | NP_004635.2 | 6 | 750 | |||
XM_017022640.1 | XP_016878129.1 | 6 | 954 | |||
XM_017022641.1 | XP_016878130.1 | 6 | 954 | |||
GenBank mRNA | AF022152.1 | - | 6 | 579 | ||
BC026700.1 | - | 6 | 702 | |||
BC093739.1 | - | 6 | 621 | 7939584 | ||
BC143342.1 | - | 5 | 525 | 9051847 | ||
BC143346.1 | - | 6 | 621 | 9051851 | ||
CR749860.1 | - | 5 | 750 | |||
U37673.1 | - | 6 | 609 |