Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkacb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164198.1 | NP_001157670.1 | 3 | 460 | ||
NM_001164199.1 | NP_001157671.1 | 3 | 333 | |||
NM_001164200.1 | NP_001157672.1 | 3 | 425 | |||
NM_011100.4 | NP_035230.1 | 3 | 464 | |||
GenBank mRNA | AK048319.1 | - | 3 | 456 | ||
AK139419.1 | - | 3 | 205 | |||
AK148728.1 | - | 3 | 274 | |||
AK150122.1 | - | 3 | 426 | |||
AK150134.1 | - | 3 | 426 | |||
AK163585.1 | - | 3 | 238 | |||
AK193506.1 | - | 3 | 207 | |||
AK204631.1 | - | 3 | 207 | |||
BC054533.1 | - | 3 | 448 | 5704796 | ||
CB723114.1 | - | 3 | 460 | |||
J02626.1 | - | 3 | 393 | |||
JN189786.1 | - | 3 | 276 | |||
JN189787.1 | - | 3 | 225 | |||
JN189788.1 | - | 3 | 200 |