Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkacb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164198.1 | NP_001157670.1 | 6 | 712 | ||
NM_001164199.1 | NP_001157671.1 | 6 | 585 | |||
NM_001164200.1 | NP_001157672.1 | 6 | 677 | |||
NM_011100.4 | NP_035230.1 | 6 | 716 | |||
GenBank mRNA | AK048319.1 | - | 6 | 708 | ||
AK139419.1 | - | 6 | 457 | |||
AK148728.1 | - | 6 | 526 | |||
AK150122.1 | - | 6 | 678 | |||
AK150134.1 | - | 6 | 678 | |||
AK163585.1 | - | 6 | 490 | |||
BC054533.1 | - | 6 | 700 | 5704796 | ||
J02626.1 | - | 6 | 645 | |||
JN189786.1 | - | 6 | 528 | |||
JN189787.1 | - | 6 | 477 | |||
JN189788.1 | - | 6 | 452 |