Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pld3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011116.1 | NP_035246.1 | 9 | 1001 | ||
GenBank mRNA | AF026124.1 | - | 9 | 1001 | ||
AK153508.1 | - | 7 | 690 | |||
BC068170.1 | - | 9 | 997 | |||
BC076586.1 | - | 9 | 1065 | 30604186 |