Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pou2f2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163554.1 | NP_001157026.1 | 9 | 684 | ||
NM_001163555.1 | NP_001157027.1 | 9 | 750 | |||
NM_001163556.1 | NP_001157028.1 | 9 | 684 | |||
NM_011138.2 | NP_035268.2 | 9 | 732 | |||
GenBank mRNA | AK156322.1 | - | 9 | 697 | ||
AY746973.1 | - | 9 | 712 | |||
AY746974.1 | - | 9 | 730 | |||
BC050258.1 | - | 9 | 697 | |||
BC104488.1 | - | 9 | 684 | 40046281 | ||
BC105647.1 | - | 9 | 684 | 40046279 | ||
BC105920.1 | - | 9 | 750 | 40046289 | ||
BC105921.1 | - | 9 | 684 | 40046286 | ||
S55236.1 | - | 2 | 189 | |||
X51961.1 | - | 1 | 70 | |||
X53654.1 | - | 892 | ||||
X57089.1 | - | 2 | 189 | |||
X57936.1 | - | 9 | 684 | |||
X57937.1 | - | 9 | 732 | |||
X57938.1 | - | 9 | 750 | |||
X57939.1 | - | 9 | 684 | |||
X57940.1 | - | 9 | 684 | |||
X57941.1 | - | 8 | 567 |