Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011157.2 | NP_035287.1 | 2 | 206 | ||
GenBank mRNA | AK087960.1 | - | 1 | 35 | ||
AK151357.1 | - | 2 | 210 | |||
AK157945.1 | - | 2 | 207 | |||
AK168124.1 | - | 2 | 209 | |||
BC037076.1 | - | 2 | 196 | 5038568 | ||
J04549.1 | - | 2 | 203 | |||
M34603.1 | - | 2 | 207 | |||
X16133.1 | - | 2 | 206 |