Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk8ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202445.1 | NP_001189374.1 | 6 | 1895 | ||
NM_001202446.1 | NP_001189375.1 | 6 | 1820 | |||
NM_011162.5 | NP_035292.2 | 6 | 1603 | |||
GenBank mRNA | AB221603.1 | - | 6 | 1466 | ||
AF003115.1 | - | 6 | 1603 | |||
AF054611.1 | - | 6 | 1603 | |||
AF109768.1 | - | 6 | 1500 | |||
AF109769.1 | - | 6 | 1563 | |||
AF109770.1 | - | 6 | 1618 | |||
AF109771.1 | - | 6 | 1464 | |||
AF332075.1 | - | 6 | 1466 | |||
AF332076.1 | - | 6 | 1466 | |||
AK147033.1 | - | 6 | 1822 | |||
AK217001.1 | - | 2 | 289 | |||
AK219726.1 | - | 2 | 77 | |||
BC072578.1 | - | 6 | 1787 | 6402485 |