Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2ak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011163.4 | NP_035293.1 | 3, 4 | 434 | ||
GenBank mRNA | AK028602.1 | - | 3, 4 | 434 | ||
BC016422.1 | - | 3, 4 | 397 | 3964935 | ||
M65029.1 | - | 3, 4 | 386 | |||
M93567.1 | - | 3, 4 | 376 |