Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl7a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011168.4 | NP_035298.1 | 4 | 383 | ||
GenBank mRNA | AF011382.1 | - | 4 | 372 | ||
AF020524.1 | - | 4 | 379 | |||
AK005455.1 | - | 4 | 384 | |||
AK028068.1 | - | 4 | 383 | |||
AK131665.1 | - | 4 | 384 | |||
BC099468.1 | - | 4 | 362 | 30789902 |