Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VNN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242350.2 | NP_001229279.1 | 5 | 1119 | ||
NM_004665.4 | NP_004656.2 | 7 | 1782 | |||
NM_078488.2 | NP_511043.1 | 8 | 1610 | |||
NR_034173.2 | 6 | 1493 | ||||
NR_034174.2 | 6 | 1253 | ||||
NR_110143.1 | 7 | 1778 | ||||
NR_110144.1 | 7 | 1570 | ||||
NR_110145.1 | 7 | 1567 | ||||
NR_110146.1 | 4 | 926 | ||||
XM_006715593.3 | XP_006715656.1 | 9 | 1757 | |||
XM_017011407.1 | XP_016866896.1 | 8 | 2393 | |||
XM_017011408.1 | XP_016866897.1 | 9 | 1762 | |||
XM_017011409.1 | XP_016866898.1 | 9 | 1655 | |||
GenBank mRNA | AB026705.1 | - | 7 | 1664 | ||
AJ132100.1 | - | 7 | 1660 | |||
AK290908.1 | - | 7 | 1678 | |||
AK292125.1 | - | 8 | 1598 | |||
BC126145.1 | - | 7 | 1744 | 8991861 | ||
BC126147.1 | - | 7 | 1744 | 8991863 |