Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011188.3 | NP_035318.1 | 2 | 363 | ||
GenBank mRNA | AK076874.1 | - | 2 | 365 | ||
U61283.1 | - | 1 | 49 |