Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011188.3 | NP_035318.1 | 6 | 732 | ||
GenBank mRNA | AK005083.1 | - | 5 | 355 | ||
AK076874.1 | - | 6 | 734 | |||
AK146919.1 | - | 5 | 373 | |||
AK153413.1 | - | 5 | 376 | |||
AK168548.1 | - | 5 | 373 | |||
AK216249.1 | - | 2 | 105 | |||
BC005462.1 | - | 5 | 382 | 3590739 |