Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psme1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011189.1 | NP_035319.1 | 6, 7 | 467 | ||
GenBank mRNA | AK008514.1 | - | 6, 7 | 442 | ||
AK019390.1 | - | 6, 7 | 495 | |||
AK150708.1 | - | 6, 7 | 511 | |||
AK150975.1 | - | 6, 7 | 510 | |||
AK182144.1 | - | 2, 3 | 159 | |||
BC088981.1 | - | 6, 7 | 487 | 5689048 | ||
D87909.1 | - | 6, 7 | 467 | |||
U60328.1 | - | 6, 7 | 386 |