Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pstpip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011193.2 | NP_035323.2 | 15 | 1644 | ||
GenBank mRNA | AK037936.1 | - | 13 | 2635 | ||
AK088150.1 | - | 14 | 2446 | |||
BC096761.1 | - | 15 | 1424 | 30040955 | ||
U87814.1 | - | 15 | 1484 |