Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pth1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083935.1 | NP_001077404.1 | 15 | 1923 | ||
NM_001083936.1 | NP_001077405.1 | 14 | 1956 | |||
NM_011199.2 | NP_035329.2 | 16 | 1925 | |||
GenBank mRNA | AK134583.1 | - | 8 | 954 | ||
BC013446.1 | - | 16 | 1902 | 4241234 | ||
BC051981.1 | - | 14 | 1827 | 6402767 | ||
X78936.1 | - | 14 | 1702 |
Set Membership: |
G-protein coupled receptor |