Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011201.3 | NP_035331.3 | 3 | 334 | ||
GenBank mRNA | AK149979.1 | - | 3 | 290 | ||
AK150322.1 | - | 3 | 290 | |||
AK151371.1 | - | 3 | 247 | |||
AK153088.1 | - | 3 | 273 | |||
AK156281.1 | - | 3 | 295 | |||
AK157431.1 | - | 3 | 335 | |||
AK165567.1 | - | 3 | 299 | |||
AK165949.1 | - | 4 | 414 | |||
AK166020.1 | - | 3 | 290 | |||
AK168255.1 | - | 3 | 272 | |||
AK168772.1 | - | 3 | 289 | |||
AK170620.1 | - | 3 | 290 | |||
AK171380.1 | - | 3 | 290 | |||
AK185938.1 | - | 2 | 41 | |||
BC010191.1 | - | 3 | 252 | 3156854 | ||
M97590.1 | - | 3 | 250 |