Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011203.2 | NP_035333.2 | 4 | 362 | ||
GenBank mRNA | AK154107.1 | - | 4 | 454 | ||
BC051980.1 | - | 4 | 362 | 6402545 | ||
X63440.1 | - | 4 | 363 |