Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabggtb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163478.1 | NP_001156950.1 | 2 | 104 | ||
NM_011231.2 | NP_035361.2 | 1 | 242 | |||
GenBank mRNA | AF231926.1 | - | 1 | 98 | ||
AK159902.1 | - | 2 | 452 | |||
AK160162.1 | - | 2 | 91 | |||
BC003948.1 | - | 1 | 85 | |||
BC021335.1 | - | 1 | 90 | |||
BC057661.1 | - | 1 | 350 | |||
BC132473.1 | - | 1 | 120 | 40130750 | ||
BC138547.1 | - | 1 | 120 | 8861567 | ||
BC145323.1 | - | 1 | 120 | |||
BC145324.1 | - | 1 | 120 | |||
BC145325.1 | - | 1 | 120 | |||
BC145326.1 | - | 1 | 120 | |||
BX512972.1 | - | 1 | 242 | |||
CF732295.1 | - | 1 | 77 | |||
CX205055.1 | - | 2 | 104 | |||
U12922.1 | - | 1 | 98 |