Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044371.2 | NP_001037836.1 | 14 | 1651 | ||
NM_001283011.1 | NP_001269940.1 | 15 | 1856 | |||
NM_011233.3 | NP_035363.2 | 14 | 1779 | |||
GenBank mRNA | AF085737.1 | - | 14 | 1702 | ||
AJ011923.1 | - | 13 | 1552 | |||
AK034773.1 | - | 1 | 53 | |||
AK134277.1 | - | 14 | 1606 | |||
AK162016.1 | - | 14 | 1639 | |||
AK167358.1 | - | 15 | 1799 | |||
BC050267.1 | - | 11 | 1291 | |||
BC066855.1 | - | 14 | 1769 | 30473984 |