Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004688.2 | NP_004679.2 | 5, 6 | 785 | ||
XM_005246941.2 | XP_005246998.1 | 5, 6 | 648 | |||
XM_017005247.1 | XP_016860736.1 | 5, 6 | 858 | |||
GenBank mRNA | AB451305.1 | - | 4, 5 | 455 | ||
AB451436.1 | - | 4, 5 | 455 | |||
AK291548.1 | - | 5, 6 | 773 | |||
AK298754.1 | - | 5 | 860 | |||
BC001268.1 | - | 5, 6 | 505 | 3452659 | ||
BC021987.2 | - | 4, 5 | 461 | 4096531 | ||
U32849.1 | - | 5, 6 | 735 |