Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasgrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011242.2 | NP_035372.2 | 3 | 249 | ||
GenBank mRNA | BC027446.1 | - | 2 | 1788 | ||
BC051474.1 | - | 2 | 208 | |||
CJ057054.1 | - | 3 | 250 | |||
CO425553.1 | - | 2 | 174 |