Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011251.3 | NP_035381.2 | 3 | 1072 | ||
NM_029169.3 | NP_083445.1 | 3 | 1343 | |||
GenBank mRNA | AJ006486.2 | - | 3 | 951 | ||
AK077899.1 | - | 3 | 1250 | |||
AK081846.1 | - | 3 | 997 | |||
AK134164.1 | - | 3 | 983 | |||
AK145612.1 | - | 3 | 1232 | |||
AK220446.1 | - | 3 | 1221 | |||
BC026129.1 | - | 3 | 1097 | 3982861 | ||
BC026130.1 | - | 3 | 1246 |