Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm21708 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270516.1 | NP_001257445.1 | 9 | 940 |
Gene Symbol: Gm10256 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270510.1 | NP_001257439.1 | 940 |
Gene Symbol: Gm10352 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270511.1 | NP_001257440.1 | 940 |
Gene Symbol: Gm21677 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270513.1 | NP_001257442.1 | 940 |
Gene Symbol: Gm21693 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270514.1 | NP_001257443.1 | 940 |
Gene Symbol: Gm21704 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270515.1 | NP_001257444.1 | 940 |
Gene Symbol: Gm3376 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270512.1 | NP_001257441.1 | 9 | 940 |
Gene Symbol: Gm4064 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270518.1 | NP_001257447.1 | 9 | 940 |
Gene Symbol: Rbmy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166384.1 | NP_001159856.1 | 9 | 940 | ||
NM_011253.2 | NP_035383.2 | 10 | 1091 | |||
GenBank mRNA | BC150670.1 | - | 921 | 9087581 | ||
U36929.1 | - | 932 | ||||
Y15131.1 | - | 10 | 1080 |