Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rest | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011263.2 | NP_035393.2 | 2 | 524 | ||
GenBank mRNA | AB024496.1 | - | 1 | 220 | ||
AB221589.1 | - | 1 | 220 | |||
AK004945.2 | - | 2 | 527 | |||
AK156514.1 | - | 2 | 510 | |||
BC096434.1 | - | 2 | 447 | |||
BC127065.1 | - | 1 | 220 | 40110015 | ||
DQ644039.1 | - | 1 | 220 | |||
JN019832.1 | - | 1 | 220 |