Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rev3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011264.3 | NP_035394.2 | 4 | 748 | ||
GenBank mRNA | AB031049.1 | - | 4 | 749 | ||
AF083464.1 | - | 5 | 481 | |||
AK032532.1 | - | 4 | 792 | |||
BC048946.1 | - | 5 | 909 |