Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rev3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011264.3 | NP_035394.2 | 4, 5 | 883 | ||
GenBank mRNA | AB031049.1 | - | 4, 5 | 884 | ||
AF083464.1 | - | 5, 6 | 616 | |||
AK032532.1 | - | 4, 5 | 927 | |||
BC048946.1 | - | 5, 6 | 1044 |