Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnaseh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286865.1 | NP_001273794.1 | 8 | 1055 | ||
NM_011275.3 | NP_035405.2 | 8 | 1055 | |||
GenBank mRNA | AF048993.1 | - | 8 | 1016 | ||
AK011680.1 | - | 7 | 970 | |||
AK088032.1 | - | 6 | 1570 | |||
BC019411.1 | - | 8 | 984 | 5133942 | ||
BG084607.2 | - | 4 | 505 | |||
BM943446.1 | - | 5 | 575 | |||
DV662103.1 | - | 2 | 273 |