Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF169300.1 | - | 3 | 339 | ||
AK019171.1 | - | 3 | 354 | |||
AK019656.1 | - | 3 | 391 | |||
AK090162.1 | - | 3 | 377 | |||
AK133924.1 | - | 3 | 439 | |||
AK145596.1 | - | 3 | 316 | |||
AK147057.1 | - | 3 | 299 | |||
AK151116.1 | - | 3 | 347 | |||
AK158987.1 | - | 3 | 356 | |||
AK159296.1 | - | 3 | 354 | |||
AK159778.1 | - | 3 | 348 | |||
AK159949.1 | - | 3 | 396 | |||
AK207872.1 | - | 3 | 277 | |||
BC003282.1 | - | 3 | 318 | 3582133 | ||
BY275961.1 | - | 3 | 329 | |||
U95141.1 | - | 1 | 29 |