Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF169300.1 | - | 8 | 1341 | ||
AK019171.1 | - | 8 | 1355 | |||
AK019656.1 | - | 8 | 1392 | |||
AK037465.1 | - | 756 | ||||
AK090162.1 | - | 8 | 1379 | |||
AK145596.1 | - | 8 | 1318 | |||
AK147057.1 | - | 8 | 1301 | |||
AK151116.1 | - | 8 | 1349 | |||
AK158987.1 | - | 8 | 1358 | |||
AK159296.1 | - | 8 | 1356 | |||
AK159778.1 | - | 8 | 1350 | |||
AK159949.1 | - | 8 | 1398 | |||
AK191020.1 | - | 1 | 281 | |||
AK194306.1 | - | 1 | 283 | |||
AK195100.1 | - | 1 | 282 | |||
AK199671.1 | - | 1 | 282 | |||
AK200323.1 | - | 1 | 282 | |||
AK203627.1 | - | 1 | 283 | |||
AK214454.1 | - | 1 | 282 | |||
BC003282.1 | - | 8 | 1320 | 3582133 | ||
BC005551.1 | - | 1 | 306 |