Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011284.3 | NP_035414.3 | 7 | 689 | ||
GenBank mRNA | AK078006.1 | - | 7 | 690 | ||
AK168680.1 | - | 7 | 689 | |||
AK169847.1 | - | 7 | 691 | |||
BC004578.1 | - | 7 | 688 | 3586727 | ||
D00812.1 | - | 7 | 661 |