Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004705.3 | NP_004696.2 | 4 | 472 | ||
NR_130898.1 | 5 | 567 | ||||
GenBank mRNA | AF007393.1 | - | 4 | 343 | ||
AF081567.1 | - | 4 | 343 | |||
AK291843.1 | - | 4 | 344 | |||
AK299594.1 | - | 2 | 103 | |||
BC117138.1 | - | 4 | 401 | 40125689 | ||
BC117140.1 | - | 4 | 401 | 40125691 | ||
BX641144.1 | - | 1 | 3943 | |||
DR005363.1 | - | 5 | 573 |