Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011289.3 | NP_035419.1 | 4, 5 | 582 | ||
GenBank mRNA | AF214527.1 | - | 4, 5 | 425 | ||
AK002945.1 | - | 4, 5 | 399 | |||
AK008120.1 | - | 4, 5 | 407 | |||
AK010627.1 | - | 4, 5 | 400 | |||
AK012441.1 | - | 2, 3 | 217 | |||
AK012566.1 | - | 4, 5 | 402 | |||
AK088211.1 | - | 4, 5 | 391 | |||
AK132027.1 | - | 4, 5 | 410 | |||
AK146291.1 | - | 4, 5 | 410 | |||
BC024366.1 | - | 4, 5 | 395 | 4986323 | ||
BC082284.1 | - | 4, 5 | 395 | 5339848 | ||
BC090395.1 | - | 4, 5 | 386 | 5062830 | ||
BQ748836.1 | - | 4, 5 | 387 |